Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14343801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14343801

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen Fc gamma RIIIA/CD16a
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Applications Flow Cytometry
Immunogen The immunogen for this antibody was CD16.
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2214
Clone CB16
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified