Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434380025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14343803

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Applications Flow Cytometry
Immunogen The immunogen for this antibody was CD16.
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Human
Antigen Fc gamma RIIIA/CD16a
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene ID (Entrez) 2214
Clone CB16
Concentration 0.5mg/mL