Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, FITC, Novus Biologicals™

$128.00 - $289.00

Specifications

Antigen Fc gamma RIIIA/CD16a
Clone CB16
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD16.
Gene ID (Entrez) 2214
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP144028

 
novus biologicals canada
NBP144028100TESTS
100 Tests Each for $289.00

NBP14402825

 
novus biologicals canada
NBP14402825TESTS
25 Tests Each for $128.00
Description & Specifications

Specifications

Antigen Fc gamma RIIIA/CD16a
Clone CB16
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD16.
Gene ID (Entrez) 2214
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry