Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, FITC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP144028100TESTS

 View all options for this product


For Research Use Only

Catalog No. NBP144028


Description & Specifications


Antigen Fc gamma RIIIA/CD16a
Applications Flow Cytometry
Clone CB16
Concentration 0.5mg/mL
Conjugate FITC
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Host Species Mouse
Immunogen The immunogen for this antibody was CD16.
Isotype IgG1
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2214