Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, Biotin, Novus Biologicals™

$128.00 - $246.00

Specifications

Antigen Fc gamma RIIIA/CD16a
Clone CB16
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD16.
Gene ID (Entrez) 2214
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14367101

 
novus biologicals canada
NBP14367101MG
0.1mg Each for $246.00

NBP14367103

 
novus biologicals canada
NBP1436710025MG
0.025mg Each for $128.00
Description & Specifications

Specifications

Antigen Fc gamma RIIIA/CD16a
Clone CB16
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD16.
Gene ID (Entrez) 2214
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry