Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP1436710025MG

 View all options for this product


For Research Use Only

Catalog No. NBP14367103


Description & Specifications


Antigen Fc gamma RIIIA/CD16a
Applications Flow Cytometry
Clone CB16
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was CD16.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2214