Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: CB16, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14367101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14367101

  • / Each
 

Description & Specifications

Specifications

Antigen Fc gamma RIIIA/CD16a
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Applications Flow Cytometry
Immunogen The immunogen for this antibody was CD16.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2214
Clone CB16
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Human