Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: 1B11, Novus Biologicals™

$254.34 - $310.66

Specifications

Antigen Fc gamma RIIIA/CD16a
Clone 1B11
Concentration 1mg/mL
Immunogen Recombinant human FCGR3A (18-208aa) purified from E. coli
Gene ID (Entrez) 2214
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23121301

 
novus biologicals canada
NBP23121301ML
0.1mL Each for $310.66

NBP23121305

 
novus biologicals canada
NBP231213005ML
0.05mL Each for $254.34
Description & Specifications

Specifications

Antigen Fc gamma RIIIA/CD16a
Clone 1B11
Concentration 1mg/mL
Immunogen Recombinant human FCGR3A (18-208aa) purified from E. coli
Gene ID (Entrez) 2214
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Western Blotting