Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RIIIA/CD16a, Clone: 1B11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23121301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23121301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Western Blotting
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Immunogen Recombinant human FCGR3A (18-208aa) purified from E. coli
Quantity 0.1mL
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Reconstitution Protein A purified
Host Species Mouse
Antigen Fc gamma RIIIA/CD16a
Clone 1B11
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Purification Method Purified
Concentration 1mg/mL
Isotype IgG2a, κ