Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RII/CD32, Clone: 6C4, APC, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen Fc gamma RII/CD32
Clone 6C4
Immunogen The immunogen for this antibody is CD32.
Gene ID (Entrez) 2213
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200212

 
novus biologicals canada
NBP200212100TESTS
100 Tests Each for $362.00

NBP20021225

 
novus biologicals canada
NBP20021225TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen Fc gamma RII/CD32
Clone 6C4
Immunogen The immunogen for this antibody is CD32.
Gene ID (Entrez) 2213
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry