Please login to your online account to display your discounted pricing

Mouse anti-Fc gamma RI/CD64, Clone: 10.1, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Fc gamma RI/CD64
Clone 10.1
Immunogen Tissue / cell preparation (Human): monocytes.
Gene ID (Entrez) 2209
Formulation Protein G purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200120

 
novus biologicals canada
NBP200120100TESTS
100 Tests Each for $252.00

NBP20012025

 
novus biologicals canada
NBP20012025TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen Fc gamma RI/CD64
Clone 10.1
Immunogen Tissue / cell preparation (Human): monocytes.
Gene ID (Entrez) 2209
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1
Conjugate FITC
Format Purified
Purification Method Purified
Applications Flow Cytometry