Please login to your online account to display your discounted pricing

Mouse anti-Fc epsilon RI, Clone: AER-37, Novus Biologicals™

$285.00 - $406.00

Specifications

Antigen Fc epsilon RI
Clone AER-37
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was FceRI alpha.
Gene ID (Entrez) 2205
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14327801

 
novus biologicals canada
NBP14327801MG
0.1mg Each for $406.00

NBP14327803

 
novus biologicals canada
NBP1432780025MG
0.025mg Each for $285.00
Description & Specifications

Specifications

Antigen Fc epsilon RI
Clone AER-37
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was FceRI alpha.
Gene ID (Entrez) 2205
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry