Please login to your online account to display your discounted pricing

Mouse anti-Fc epsilon RI, Clone: AER-37, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327801

  • / Each
 

Description & Specifications

Specifications

Immunogen The immunogen for this antibody was FceRI alpha.
Applications Flow Cytometry
Format Affinity Purified
Cross Reactivity Human
Storage Requirements Store at 4°C. Do not freeze.
Antigen Fc epsilon RI
Quantity 0.1mg
Primary or Secondary Primary
Regulatory Status RUO
Isotype IgG2b, κ
Host Species Mouse
Concentration 0.5mg/mL
Clone AER-37
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Purification Method Affinity Purified
Gene ID (Entrez) 2205