Please login to your online account to display your discounted pricing

Mouse anti-Fc epsilon RI, Clone: AER-37, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen Fc epsilon RI
Clone AER-37
Immunogen The immunogen for this antibody was FceRI alpha.
Gene ID (Entrez) 2205
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14352201

 
novus biologicals canada
NBP14352201MG
0.1mg Each for $329.00

NBP14352203

 
novus biologicals canada
NBP1435220025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen Fc epsilon RI
Clone AER-37
Immunogen The immunogen for this antibody was FceRI alpha.
Gene ID (Entrez) 2205
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry