Please login to your online account to display your discounted pricing

Mouse anti-Fas/TNFRSF6/CD95, Clone: 15A7, Novus Biologicals™

$274.00 - $362.00

Specifications

Antigen Fas/TNFRSF6/CD95
Clone 15A7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD95.
Gene ID (Entrez) 355
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1433775

 
novus biologicals canada
NBP14337705MG
0.5mg Each for $362.00

NBP1433775M

 
novus biologicals canada
NBP143377005MG
0.05mg Each for $274.00
Description & Specifications

Specifications

Antigen Fas/TNFRSF6/CD95
Clone 15A7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD95.
Gene ID (Entrez) 355
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry