Please login to your online account to display your discounted pricing

Mouse anti-Fas/TNFRSF6/CD95, Clone: 15A7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14337705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433775

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Quantity 0.5mg
Immunogen The immunogen for this antibody was CD95.
Antigen Fas/TNFRSF6/CD95
Host Species Mouse
Concentration 0.5mg/mL
Cross Reactivity Mouse
Gene ID (Entrez) 355
Formulation Phosphate buffer pH 7.2
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Clone 15A7
Purification Method Affinity Purified
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG1, κ