Please login to your online account to display your discounted pricing

Mouse anti-Fas/TNFRSF6/CD95, Clone: 15A7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143377005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433775M

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Quantity 0.05mg
Antigen Fas/TNFRSF6/CD95
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was CD95.
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 355
Conjugate Unlabeled
Isotype IgG1, κ
Regulatory Status RUO
Clone 15A7
Concentration 0.5mg/mL
Cross Reactivity Mouse