Please login to your online account to display your discounted pricing

Mouse anti-FABP9/T-FABP, Clone: 13F9, Novus Biologicals™

$254.34 - $310.66

Specifications

Antigen FABP9/T-FABP
Clone 13F9
Concentration 1mg/mL
Immunogen Recombinant human FABP9 (1-132aa) purified from E. coli
Gene Accession No. NP_001073995
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23120001

 
novus biologicals canada
NBP23120001ML
0.1mL Each for $310.66

NBP23120005

 
novus biologicals canada
NBP231200005ML
0.05mL Each for $254.34
Description & Specifications

Specifications

Antigen FABP9/T-FABP
Clone 13F9
Concentration 1mg/mL
Immunogen Recombinant human FABP9 (1-132aa) purified from E. coli
Gene Accession No. NP_001073995
Gene ID (Entrez) 646480
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Western Blot