Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: SPM567, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Estrogen Receptor
Clone SPM567
Concentration 0.2mg/mL
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Gene Accession No. P03372
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23296501

 
novus biologicals canada
NBP23296501ML
0.1mL Each for $139.00

NBP23296510

 
novus biologicals canada
NBP23296510ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Estrogen Receptor
Clone SPM567
Concentration 0.2mg/mL
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Gene Accession No. P03372
Gene ID (Entrez) 2099
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting