Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: SPM567, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23296510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23296510

  • / Each
 

Description & Specifications

Specifications

Antigen Estrogen Receptor
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM567
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P03372
Host Species Mouse
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2099
Reconstitution Protein A purified