Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: ER506, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Estrogen Receptor
Clone ER506
Concentration 1.0mg/mL
Immunogen Within the range of amino acids 2-197 of human Estrogen Receptor alpha were used as immunogen for this antibody.
Gene Accession No. P03372
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23332101

 
novus biologicals canada
NBP23332101MG
0.1mg Each for $384.00

NBP23332102

 
novus biologicals canada
NBP23332102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Estrogen Receptor
Clone ER506
Concentration 1.0mg/mL
Immunogen Within the range of amino acids 2-197 of human Estrogen Receptor alpha were used as immunogen for this antibody.
Gene Accession No. P03372
Gene ID (Entrez) 2099
Formulation PBS. Does not contain BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot