Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen Estrogen Receptor
Clone ER503
Concentration 0.2mg/mL
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Gene Accession No. P03372
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23332201

 
novus biologicals canada
NBP23332201MG
0.1mg Each for $329.00

NBP23332202

 
novus biologicals canada
NBP23332202MG
0.2mg Each for $472.00
Description & Specifications

Specifications

Antigen Estrogen Receptor
Clone ER503
Concentration 0.2mg/mL
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Gene Accession No. P03372
Gene ID (Entrez) 2099
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot