Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23332202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23332202

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.2mg
Primary or Secondary Primary
Antigen Estrogen Receptor
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P03372
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2099
Clone ER503
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)