Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23332201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23332201

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone ER503
Cross Reactivity Human
Antigen Estrogen Receptor
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P03372
Host Species Mouse
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Isotype IgG1
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 2099
Concentration 0.2mg/mL
Conjugate Unlabeled