Please login to your online account to display your discounted pricing

Mouse anti-Estrogen Receptor, Clone: ER503, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23296601ML

 View all options for this product


For Research Use Only

Catalog No. NBP23296601


Description & Specifications


Antigen Estrogen Receptor
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone ER503
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P03372
Host Species Mouse
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2099