Please login to your online account to display your discounted pricing

Mouse anti-Erythrocyte Specific, Clone: SFL23.6, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471501

  • / Each
 

Description & Specifications

Specifications

Antigen Erythrocyte Specific
Host Species Mouse
Cross Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Immunogen Hepatocytes from 20-22 week-old human fetus
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SFL23.6
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified