missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-ErbB2/Her2, Clone: HRB2/282, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen ErbB2/Her2
Clone HRB2/282
Concentration 1mg/mL
Immunogen Recombinant human HER-2 protein
Gene Accession No. P04626
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23464201 Novus Biologicals Canada
NBP23464201MG
0.1mg Each for $384.00
NBP23464202 Novus Biologicals Canada
NBP23464202MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

ErbB2/Her2
1mg/mL
P04626
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
HRB2/282
Recombinant human HER-2 protein
2064
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation