Please login to your online account to display your discounted pricing

Mouse anti-ErbB2/Her2, Clone: HRB2/282, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Recombinant human HER-2 protein
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen ErbB2/Her2
Gene Accession No. P04626
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 2064
Reconstitution Protein A purified
Clone HRB2/282
Concentration 1mg/mL
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.