Please login to your online account to display your discounted pricing

Mouse anti-ErbB2/Her2, Clone: HRB2/258, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen ErbB2/Her2
Clone HRB2/258
Concentration 1mg/mL
Immunogen Recombinant human HER-2 protein
Gene Accession No. P04626
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23464101

 
novus biologicals canada
NBP23464101MG
0.1mg Each for $384.00

NBP23464102

 
novus biologicals canada
NBP23464102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen ErbB2/Her2
Clone HRB2/258
Concentration 1mg/mL
Immunogen Recombinant human HER-2 protein
Gene Accession No. P04626
Gene ID (Entrez) 2064
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting