Please login to your online account to display your discounted pricing

Mouse anti-ErbB2/Her2, Clone: HRB2/258, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Immunogen Recombinant human HER-2 protein
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Rat
Regulatory Status RUO
Format Purified
Isotype IgG2a, κ
Antigen ErbB2/Her2
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Reconstitution Protein A purified
Clone HRB2/258
Gene ID (Entrez) 2064
Gene Accession No. P04626
Concentration 1mg/mL
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.