Please login to your online account to display your discounted pricing

Mouse anti-ErbB2/Her2, Clone: HRB2/258, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464102

  • / Each
 

Description & Specifications

Specifications

Antigen ErbB2/Her2
Format Purified
Gene Accession No. P04626
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 2064
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Recombinant human HER-2 protein
Isotype IgG2a, κ
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone HRB2/258
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Rat
Purification Method Purified