Please login to your online account to display your discounted pricing

Mouse anti-ER alpha/NR3A1, Clone: SPM567, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen ER alpha/NR3A1
Clone SPM567
Concentration 1mg/mL
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Gene Accession No. P03372
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23480101

 
novus biologicals canada
NBP23480101MG
0.1mg Each for $384.00

NBP23480102

 
novus biologicals canada
NBP23480102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen ER alpha/NR3A1
Clone SPM567
Concentration 1mg/mL
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Gene Accession No. P03372
Gene ID (Entrez) 2099
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blot