Please login to your online account to display your discounted pricing

Mouse anti-ER alpha/NR3A1, Clone: SPM567, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480102

  • / Each
 

Description & Specifications

Specifications

Antigen ER alpha/NR3A1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone SPM567
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P03372
Host Species Murine
Immunogen Recombinant human Estrogen Receptor alpha protein (aa2-185)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2099
Test Specificity This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family.
Reconstitution Protein A purified
Target Species Human