missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-EPX, Clone: EPO104, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen EPX
Clone EPO104
Concentration 1mg/mL
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Gene Accession No. P11678
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23461401 Novus Biologicals Canada
NBP23461401MG
0.1mg Each for $384.00
NBP23461402 Novus Biologicals Canada
NBP23461402MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

EPX
1mg/mL
P11678
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
EPO104
Human eosinophils from a patient with hypereosinophilic syndrome
8288
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen)