Please login to your online account to display your discounted pricing

Mouse anti-EPX, Clone: EPO104, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461402

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Antigen EPX
Host Species Mouse
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Monoclonal or Polyclonal Monoclonal
Clone EPO104
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P11678
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 8288
Reconstitution Protein A purified
Concentration 1mg/mL
Quantity 0.2mg