Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM535, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen EpCAM/TROP1
Clone SPM535
Immunogen Rat small intestine cells
Gene ID (Entrez) 4072
Formulation Culture Medium. Bioreactor concentrate.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23445601

 
novus biologicals canada
NBP23445601ML
0.1mL Each for $139.00

NBP23445605

 
novus biologicals canada
NBP23445605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen EpCAM/TROP1
Clone SPM535
Immunogen Rat small intestine cells
Gene ID (Entrez) 4072
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting