Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM535, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23445605ML

 View all options for this product


For Research Use Only

Catalog No. NBP23445605


Description & Specifications


Antigen EpCAM/TROP1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM535
Conjugate Unlabeled
Cross Reactivity Rat
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Immunogen Rat small intestine cells
Isotype IgG1
Purification Method Concentrated
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity Does not react to Human.