Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM534, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23445501ML

 View all options for this product


For Research Use Only

Catalog No. NBP23445501


Description & Specifications


Antigen EpCAM/TROP1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM534
Conjugate Unlabeled
Cross Reactivity Rat
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Immunogen Rat small intestine cells
Isotype IgG1
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity Does not react to Human.