Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM534, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445505

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Rat
Antigen EpCAM/TROP1
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Rat small intestine cells
Isotype IgG1
Purification Method Concentrated
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity Does not react to Human.
Conjugate Unlabeled
Quantity 0.5mL
Clone SPM534