Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM528, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen EpCAM/TROP1
Clone SPM528
Immunogen Neuraminidase treated GLS-1 human small cell lung carcinoma cells
Gene Accession No. P16422
Gene ID (Entrez) 4072
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23445401

 
novus biologicals canada
NBP23445401ML
0.1mL Each for $139.00

NBP23445405

 
novus biologicals canada
NBP23445405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen EpCAM/TROP1
Clone SPM528
Immunogen Neuraminidase treated GLS-1 human small cell lung carcinoma cells
Gene Accession No. P16422
Gene ID (Entrez) 4072
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting