Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM528, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445405

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Antigen EpCAM/TROP1
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P16422
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Concentrated
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone SPM528
Immunogen Neuraminidase treated GLS-1 human small cell lung carcinoma cells
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity Binding epitope of this antibody is located in the first EGF-like repeat domain (EGF1) between amino acids 27-59 of Ep-CAM. Does not react with Rat.
Cross Reactivity Human