Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM528, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 4072
Immunogen Neuraminidase treated GLS-1 human small cell lung carcinoma cells
Primary or Secondary Primary
Quantity 0.1mL
Conjugate Unlabeled
Format Concentrated
Cross Reactivity Human
Test Specificity Binding epitope of this antibody is located in the first EGF-like repeat domain (EGF1) between amino acids 27-59 of Ep-CAM. Does not react with Rat.
Antigen EpCAM/TROP1
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Purification Method Concentrated
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Clone SPM528
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P16422
Host Species Mouse