Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: SPM134, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439801

  • / Each
 

Description & Specifications

Specifications

Antigen EpCAM/TROP1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM134
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P16422
Host Species Murine
Immunogen Small cell lung carcinoma cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma. Does not react with Rat.
Reconstitution Protein A purified
Target Species Human