Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: MOC-31, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP22238805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP22238805

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Primary or Secondary Primary
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 4072
Immunogen 1984).
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Clone MOC-31
Host Species Mouse
Regulatory Status RUO
Gene Accession No. AAH94465.1
Conjugate Unlabeled
Purification Method Concentrated
Test Specificity Binding epitope of this antibody is located in the first EGF-like repeat domain (EGF1) between amino acids 27-59 of Ep-CAM. EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma. Does not cross react to Rat.
Antigen EpCAM/TROP1
Quantity 0.5mL
Formulation Culture Medium. Bioreactor concentrate.
Format Concentrated
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.