Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 37F9, Novus Biologicals™

$254.67 - $310.66

Specifications

Antigen EpCAM/TROP1
Clone 37F9
Concentration 1mg/mL
Immunogen Recombinant human EPCAM (24-265aa) purified from E. coli
Gene Accession No. NP_002345
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23120401

 
novus biologicals canada
NBP23120401ML
0.1mL Each for $310.66

NBP23120405

 
novus biologicals canada
NBP231204005ML
0.05mL Each for $254.67
Description & Specifications

Specifications

Antigen EpCAM/TROP1
Clone 37F9
Concentration 1mg/mL
Immunogen Recombinant human EPCAM (24-265aa) purified from E. coli
Gene Accession No. NP_002345
Gene ID (Entrez) 4072
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Western Blotting