Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 37F9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23120401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23120401

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Antigen EpCAM/TROP1
Applications ELISA
Applications Western Blotting
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Clone 37F9
Concentration 1mg/mL
Cross Reactivity Human
Format Purified
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Gene Accession No. NP_002345
Immunogen Recombinant human EPCAM (24-265aa) purified from E. coli
Purification Method Purified
Conjugate Unlabeled
Host Species Mouse
Isotype IgG1, κ
Test Specificity Anti-human EPCAM mAb, clone AT37F9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human EPCAM protein.
Reconstitution Protein A purified