Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 323/A3, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen EpCAM/TROP1
Clone 323/A3
Concentration 0.2mg/mL
Immunogen MCF-7 human breast cancer cells
Gene Accession No. P16422
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23427401

 
novus biologicals canada
NBP23427401ML
0.1mL Each for $139.00

NBP23427405

 
novus biologicals canada
NBP23427405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen EpCAM/TROP1
Clone 323/A3
Concentration 0.2mg/mL
Immunogen MCF-7 human breast cancer cells
Gene Accession No. P16422
Gene ID (Entrez) 4072
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting