Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 323/A3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427405

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen EpCAM/TROP1
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Gene Accession No. P16422
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Clone 323/A3
Format Purified
Immunogen MCF-7 human breast cancer cells
Test Specificity Does not react to Rat.
Reconstitution Protein A purified
Concentration 0.2mg/mL