Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 323/A3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23427401ML

 View all options for this product


For Research Use Only

Catalog No. NBP23427401


Description & Specifications


Antigen EpCAM/TROP1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 323/A3
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P16422
Host Species Mouse
Immunogen MCF-7 human breast cancer cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4072
Test Specificity Does not react to Rat.
Reconstitution Protein A purified