Please login to your online account to display your discounted pricing

Mouse anti-EpCAM/TROP1, Clone: 323/A3, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen EpCAM/TROP1
Clone 323/A3
Concentration 1mg/mL
Immunogen MCF-7 human breast cancer cells
Gene Accession No. P16422
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23453202

 
novus biologicals canada
NBP23453202MG
0.2mg Each for $527.00

NBP23453201

 
novus biologicals canada
NBP23453201MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen EpCAM/TROP1
Clone 323/A3
Concentration 1mg/mL
Immunogen MCF-7 human breast cancer cells
Gene Accession No. P16422
Gene ID (Entrez) 4072
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting